Thermo Scientific Chemicals

Rhenium(VI) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 062109.14
25 g, Each
Thermo Scientific Chemicals

Rhenium(VI) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 062109.14
25 g, Each
Quantity
Catalog number: 062109.14
also known as 062109-14
Price (USD)
Price: 2,117.00
Online price: 1,799.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1314-28-9
IUPAC Name
trioxorhenium
Molecular Formula
O3Re
InChI Key
YSZJKUDBYALHQE-UHFFFAOYSA-N
SMILES
O=[Re](=O)=O
Appearance (Color)
Dark red
Total Metal Impurities
≤0.1% (1000 ppm)
Form
Powder

Description

Rhenium(VI) oxide is involved as a catalyst for the hydrogenation of carboxylic acids and carboxamides. It is involved in the alkene epoxidation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text