Thermo Scientific Chemicals

Selenium dichloride oxide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 068108.AB
10 mL, Each
Thermo Scientific Chemicals

Selenium dichloride oxide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 068108.AB
10 mL, Each
Quantity
Catalog number: 068108.AB
also known as 068108-AB
Price (USD)
Price: 336.00
Online price: 285.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7791-23-3
IUPAC Name
selenooyl dichloride
Molecular Formula
Cl2OSe
InChI Key
LIRMPIXWGWLNND-UHFFFAOYSA-N
SMILES
Cl[Se](Cl)=O
Total Metal Impurities
1% max.

Description

Selenium(IV) chloride oxide is used as a polar solvent for synthetic phenolic resins. It is also used to prepare selenium oxybromide and selenium nitride. It acts as a chlorinating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text