Thermo Scientific Chemicals

Sodium tellurate(VI) hydrate, Te 45% min, Thermo Scientific Chemicals

Catalog number: 075106.09
10 g, Each
Thermo Scientific Chemicals

Sodium tellurate(VI) hydrate, Te 45% min, Thermo Scientific Chemicals

Catalog number: 075106.09
10 g, Each
Quantity
Catalog number: 075106.09
also known as 075106-09
Price (USD)
Price: 70.00
Online price: 59.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10101-83-4
IUPAC Name
disodium tellurate
Molecular Formula
Na2O4Te
InChI Key
XERQTZLDFHNZIC-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-][Te]([O-])(=O)=O
Assay (unspecified)
Te 45% min

Description

Sodium tellurite is an intermediate in the extraction of the element, tellurium; it is a product obtained from anode slimes and is a precursor to tellurium. Solutions of sodium tellurite are used for black or blue-black coatings on iron, steel, aluminum, and copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text