Titanium(II) oxide, 99.5% (metals basis)
Titanium(II) oxide, 99.5% (metals basis)
Titanium(II) oxide, 99.5% (metals basis)
Thermo Scientific Chemicals

Titanium(II) oxide, 99.5% (metals basis)

CAS: 12137-20-1 | OTi | 63.87 g/mol
数量:
5 g
25 g
100 g
500 g
製品番号(カタログ番号) 077126.14
または、製品番号077126-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS12137-20-1
IUPAC Nameoxotitanium
Molecular FormulaOTi
InChI KeyOGIDPMRJRNCKJF-UHFFFAOYSA-N
SMILESO=[Ti]
さらに表示
Total Metal Impurities0.5% max.
Titanium(II) oxide associated with vanadium oxide forms a transition-metal-oxide monolayer catalyst, which is used in the selective oxidation and ammoxidation of alkylaromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only