Titanium(IV) oxide bis(2,4-pentanedionate)
Titanium(IV) oxide bis(2,4-pentanedionate)
Titanium(IV) oxide bis(2,4-pentanedionate)
Thermo Scientific Chemicals

Titanium(IV) oxide bis(2,4-pentanedionate)

CAS: 14024-64-7 | C10H16O5Ti | 264.1 g/mol
数量:
10 g
50 g
250 g
製品番号(カタログ番号) 077130.30
または、製品番号077130-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS14024-64-7
IUPAC Namebis(4-hydroxypent-3-en-2-one); oxotitanium
Molecular FormulaC10H16O5Ti
InChI KeyADVORQMAWLEPOI-UHFFFAOYSA-N
SMILESO=[Ti].CC(O)=CC(C)=O.CC(O)=CC(C)=O
さらに表示
Assay from Supplier's CofA≥98.0% (titration)
FormCrystals or crystalline powder
Appearance (Color)Pale yellow
Employed as a cross linking agent for cellulose fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only