Thermo Scientific Chemicals

Niobium(V) fluoride, 99%, Thermo Scientific Chemicals

Catalog number: 087221.18
50 g, Each
Thermo Scientific Chemicals

Niobium(V) fluoride, 99%, Thermo Scientific Chemicals

Catalog number: 087221.18
50 g, Each
Quantity
Catalog number: 087221.18
also known as 087221-18
Price (USD)
Price: 499.00
Online price: 432.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7783-68-8
IUPAC Name
niobium(5+) pentafluoride
Molecular Formula
F5Nb
InChI Key
AOLPZAHRYHXPLR-UHFFFAOYSA-I
SMILES
[F-].[F-].[F-].[F-].[F-].[Nb+5]
Assay (unspecified)
98.6% min

Description

Niobium(V) fluoride is used as a starting material to prepare other niobium compounds. It acts as an intermediate during the recovery of niobium metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text