Sodium cyanoborohydride, 95%
Sodium cyanoborohydride, 95%
Sodium cyanoborohydride, 95%
Thermo Scientific Chemicals

Sodium cyanoborohydride, 95%

Selective Reducing Agent For Aldehydes, Ketones, Oximes | CAS: 25895-60-7 | CH3BNNa | 62.84 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) 087839.14
または、製品番号087839-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS25895-60-7
IUPAC Namesodium cyanoboranuide
Molecular FormulaCH3BNNa
InChI KeyCVDUGUOQTVTBJH-UHFFFAOYSA-N
SMILES[Na+].[BH3-]C#N
さらに表示
Appearance (Color)White to pale cream or pale yellow
Assay from Supplier's CofA≥94.0 to ≤106.0% (Titration)
FormPowder
Selective reducing agent for aldehydes, ketones, oximes, enamines. Reductive animation of ketones, aldehydes. Reductive alkylation of amines, hydrazines Sodium cyanoborohydride is used as a reagent involved in the reductive amination of aldehydes, ketones, oximes and enamines. It is also used in the reductive alkylation of amines and hydrazines. It acts as a catalyst in the preparation of phenolate-bridged dilanthanum(III) complex. In addition, it is used in the conversion of imines to amines. It plays an important role in the reductive deoxygenation of ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Moisture Sensitive; Highly flammable
RUO – Research Use Only