Ammonium hydroxide, 25% NH{3}, 99.99% (metals basis)
Ammonium hydroxide, 25% NH{3}, 99.99% (metals basis)
Ammonium hydroxide, 25% NH{3}, 99.99% (metals basis)
Thermo Scientific Chemicals

Ammonium hydroxide, 25% NH{3}, 99.99% (metals basis)

CAS: 1336-21-6 | H5NO | 35.05 g/mol
数量:
250 mL
1000 mL
製品番号(カタログ番号) 087903.AU
または、製品番号087903-AU
価格(JPY)
-
数量:
1000 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1336-21-6
IUPAC Nameamine hydrate
Molecular FormulaH5NO
InChI KeyVHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILESN.O
さらに表示
Total Metal Impurities0.01% max.
Ammonium hydroxide is widely utilized as a leavening agent or acidity regulator in food production. It serves as a precursor to some alkyl amines and is also used in the tobacco industry for flavor enhancement and as a processing aid. During furniture making, it combines with tannic acid and is used to darken or stain wood by making it iron salts. In chemical laboratories, it used for qualitative inorganic analysis, as a complexant and as a base. It is used to clean gold, silver and platinum jewelry. It is an active component of Tollens' reagent (consisting of a solution of silver nitrate and ammonia) and is used to determine the presence of aldehyde or alpha-hydroxy ketone functional groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only