Zinc nitride, 99% (metals basis)
Zinc nitride, 99% (metals basis)
Zinc nitride, 99% (metals basis)
Thermo Scientific Chemicals

Zinc nitride, 99% (metals basis)

CAS: 1313-49-1 | N2Zn3 | 224.15 g/mol
製品番号(カタログ番号) 088212.04
または、製品番号088212-04
価格(JPY)
-
数量:
2 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1313-49-1
IUPAC Namezinc(2+) bis(zincioazanide)
Molecular FormulaN2Zn3
InChI KeyAKJVMGQSGCSQBU-UHFFFAOYSA-N
SMILES[Zn++].[N-]=[Zn].[N-]=[Zn]
さらに表示
Total Metal Impurities1% max.
Zinc nitride is useful in optical and electronic devices. It is doped with oxygen to prepare a potential nitride-based transparent conductor, which is used in semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only