Sodium hexafluoroaluminate, 97.8+%, Thermo Scientific Chemicals
Sodium hexafluoroaluminate, 97.8+%, Thermo Scientific Chemicals
Sodium hexafluoroaluminate, 97.8+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium hexafluoroaluminate, 97.8+%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
088275.36
または、製品番号088275-36
500 g
製品番号(カタログ番号) 088275.36
または、製品番号088275-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13775-53-6
IUPAC Namealuminium(3+) trisodium hexafluoride
Molecular FormulaAlF6Na3
InChI KeyREHXRBDMVPYGJX-UHFFFAOYSA-H
SMILES[F-].[F-].[F-].[F-].[F-].[F-].[Na+].[Na+].[Na+].[Al+3]
さらに表示
Assay (unspecified)97.8% min
Sodium hexafluoroaluminate is a biochemical for proteomics research. It is used as a flux agent or bath material by aluminum smelters in the production of aluminum. It is also used in the production of synthetic resins for abrasives and in the manufacture of cutting or grinding discs. Minor uses are as a coloring agent (opacifier) in the glass, ceramic and pyrotechnic industries. It occurs naturally as the mineral cryolite, which is used extensively in the industrial production of aluminum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only