Stainless Steel powder, -325 mesh, Type 316-L
Stainless Steel powder, -325 mesh, Type 316-L
Stainless Steel powder, -325 mesh, Type 316-L
Thermo Scientific Chemicals
Stainless Steel powder, -325 mesh, Type 316-L
数量:
100 g
500 g
2 kg
製品番号(カタログ番号) 088390.36
または、製品番号088390-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
仕様
平均モル重量またはモル重量範囲Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
化学物質名または材質Stainless Steel powder, Type 316-L
Gray
健康被害1H317-H350-H372
健康被害2GHS H Statement
H372-H351-H319-H317-H335
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
さらに表示
Stainless Steel powder is employed in pharmaceuticals, marine applications, architectural applications, medical implants, including pins, screws and orthopaedic implants like total hip and knee replacements.