Thermo Scientific Chemicals

Borane-d{3}, 1M in tetrahydrofuran, Thermo Scientific Chemicals

Catalog number: 089121.AD
50 mL, Each
Thermo Scientific Chemicals

Borane-d{3}, 1M in tetrahydrofuran, Thermo Scientific Chemicals

Catalog number: 089121.AD
50 mL, Each
Quantity
Catalog number: 089121.AD
also known as 089121-AD
Price (USD)

Chemical Identifiers

CAS
13763-62-7
IUPAC Name
(²H₃)borane
Molecular Formula
BD3
InChI Key
UORVGPXVDQYIDP-FIBGUPNXSA-N
SMILES
[2H]B([2H])[2H]
Assay from Supplier's CofA
≥98.0% ²H nmr
Assay from Supplier's CofA
≥98.0% Isotopic enrichment by GCMS
Stabilizer
(0.005M NaBD₄)
Appearance (Color)
Hazy
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text