Fiberlite F6-10 x 1000 LEX Carbon Fiber Rotor (for BIOS A Large-Capacity Centrifuge)
Thermo Scientific™

Fiberlite F6-10 x 1000 LEX Carbon Fiber Rotor (for BIOS A Large-Capacity Centrifuge)

Accelerate bioprocessing with the 10-liter capacity and an RCF of up to 12,000 x G of the Thermo Scientific™ Fiberlite™ F6-10x1000 LEX rotor.
Have Questions?
Catalog NumberCapacity
096-10100110 x 1000 mL
Catalog number 096-101001
Price (USD)
46,495.00
Online exclusive
54,700.00
Save 8,205.00
Each
Capacity:
10 x 1000 mL
Price (USD)
46,495.00
Online exclusive
54,700.00
Save 8,205.00
Each
Accelerate bioprocessing with the 10-liter capacity and an RCF of up to 12,000 x G of the Thermo Scientific™ Fiberlite™ F6-10x1000 LEX rotor.

Batch biprocessing with the Thermo Scientific™ Sorvall™ RC BIOS A centrifuge system.

High throughput reduces cycle times by spinning up to 10 x 1 liter bottles in a single spin.

Enhance productivity with Fiberlite carbon fiber rotors

  • Unequalled structural integrity:
    • Fatigue resistance eliminates the need to derate or limit the speed over the rotor lifespan
    • Corrosion-resistant design protects against ever-present hazard due to moisture, chemicals, alkaline solutions or other salts in the laboratory
  • Load tubes directly into the rotor, eliminating multi-canister assemblies, which can be misplaced or damaged
  • Secure investment, backed by a 15-year warranty

Ordering information: Order rotor body (096-01001) for complete assembly. Rotor Body comes with a set of Thermo Scientific Fiberlite Superspeed PPCO bottles 010-1491. For rotor care kit, order 021-101001; Lid assembly, order 099-101001;  Contact your sales representative for information.

Compatible with: Thermo Scientific™ Sorvall™ RC BIOS A, Thermo Scientific™ Fiberlite™ superspeed bottles (010-1491 and 0101-1492)

Specifications
Angle25°
AutoclavableAutoclavable
Avg. RCF12,000 x g
DescriptionFiberlite F6-10x1000 LEX Carbon Fiber Rotor
For Use With (Equipment)Sorvall BIOS A Centrifuge Heavy Duty
IncludesComplete with set of 10 bottles
K Factor5135
Max. Tube Diameter98 mm
Max. Tube Length195 mm
Net Weight31.7 lb.
No. of Places10
Radius (Metric)274 mm
Radius (Metric) Min.124 mm
BiocontainmentNo
Capacity10 x 1000 mL
MaterialCarbon Fiber
Max. RCF12,000 x g
Max. Speed6,250 rpm
TypeRotor
Unit SizeEach