Product 100052500 is equivalent Halogen Lamp to Cat. no. 4345287.
Product Image
Applied Biosystems™

Halogen Lamp for Applied Biosystems Real-Time PCR Systems

This halogen lamp is an accessory for the Applied Biosystems 7300, 7500, 7500 Fast, ViiA7, QuantStudio 6 Flex, and QuantStudioRead more
Have Questions?
Catalog NumberQuantity
1000525001 each
Catalog number 100052500
Price (USD)
160.00
Each
Quantity:
1 each
Price (USD)
160.00
Each

This halogen lamp is an accessory for the Applied Biosystems 7300, 7500, 7500 Fast, ViiA7, QuantStudio 6 Flex, and QuantStudio 7 Flex real-time PCR systems.

For Research Use Only. Not for use in diagnostics procedures.

Specifications
For Use With (Equipment)7300, 7500, 7500 Fast, ViiA7, QuantStudio 6 Flex, QuantStudio 7 Flex Systems
Quantity1 each
Light SourceHalogen
Product TypeHalogen Lamp
Unit SizeEach
Contents & Storage
Store at room temperature

Frequently asked questions (FAQs)

Can I change the lamp in the ViiA7 Real-Time PCR System by myself or does it have to be performed by a field service engineer?

You can change the lamp in the ViiA7 Real-Time PCR System without the help of an engineer. Please follow the instructions provided in Appendix B of the following maintenance guide, https://tools.thermofisher.com/content/sfs/manuals/4489821.pdf. Once the lamp has been changed, you will need to calibrate the instrument again before performing a run.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.