CytKick™ Max Tube Cooling Rack
Invitrogen™
CytKick™ Max Tube Cooling Rack
The CytKick Max Tube Cooling Rack is designed for use with the CytKick Max Autosampler and the Attune Acoustic FocusingRead more
Have Questions?
Catalog NumberQuantity
1000768231 Rack
Catalog number 100076823
Price (KRW)
2,290,000
Each
Quantity:
1 Rack
Price (KRW)
2,290,000
Each
The CytKick Max Tube Cooling Rack is designed for use with the CytKick Max Autosampler and the Attune Acoustic Focusing Cytometer. It allows sample uptake from 16 1.5- and 2.0-mL Eppendorf tubes without the need to remove the caps. This cooling rack maintains samples at 2-15°C for 108-120 minutes while they are being analyzed on the instrument. The rack is chilled by placing it in a refrigerator overnight prior to use. For a non-cooling version of this rack, please see the CytKick Max Tube Rack.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineCytKick
Quantity1 Rack
Shipping ConditionRoom Temperature
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeTube Cooling Rack
Unit SizeEach