4-Aminobutyraldehyde diethyl acetal, 95%
4-Aminobutyraldehyde diethyl acetal, 95%
4-Aminobutyraldehyde diethyl acetal, 95%
Thermo Scientific Chemicals

4-Aminobutyraldehyde diethyl acetal, 95%

CAS: 6346-09-4 | C8H19NO2 | 161.24 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 103260250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6346-09-4
IUPAC Name4,4-diethoxybutan-1-amine
Molecular FormulaC8H19NO2
InChI KeyGFLPSABXBDCMCN-UHFFFAOYSA-N
SMILESCCOC(CCCN)OCC
さらに表示
Refractive index1.4270 to 1.4310 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=94 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only