5-Aminolevulinic acid hydrochloride, 98+%
5-Aminolevulinic acid hydrochloride, 98+%
5-Aminolevulinic acid hydrochloride, 98+%
Thermo Scientific Chemicals

5-Aminolevulinic acid hydrochloride, 98+%

CAS: 5451-09-2 | C5H9NO3·HCl | 167.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
5 g
Catalog number 103920050
Price (USD)
500.65
Online exclusive
556.00
Save 55.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
500.65
Online exclusive
556.00
Save 55.35
Each
Chemical Identifiers
CAS5451-09-2
IUPAC Namehydrogen 5-amino-4-oxopentanoic acid chloride
Molecular FormulaC5H10ClNO3
InChI KeyZLHFONARZHCSET-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCC(=O)CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<0.5 %
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder or crystals
Titration Argentometric>=98.0 to 102.0 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only