2-Benzoylbenzoic Acid, 98+%
2-Benzoylbenzoic Acid, 98+%
2-Benzoylbenzoic Acid, 98+%
Thermo Scientific Chemicals

2-Benzoylbenzoic Acid, 98+%

CAS: 85-52-9 | C14H10O3 | 226.23 g/mol
数量:
25 g
500 g
1 kg
パッケージ:
Plastic bottle
製品番号(カタログ番号) 105715000
価格(JPY)
-
数量:
500 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS85-52-9
Titration with NaOH>=98.0 %
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point126°C to 130°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only