5-Benzyloxytryptamine hydrochloride, 98%
5-Benzyloxytryptamine hydrochloride, 98%
5-Benzyloxytryptamine hydrochloride, 98%
Thermo Scientific Chemicals

5-Benzyloxytryptamine hydrochloride, 98%

CAS: 52055-23-9 | C17H18N2O·HCl | 302.8 g/mol
数量:
250 mg
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 106110050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS52055-23-9
IUPAC Name2-[5-(benzyloxy)-1H-indol-3-yl]ethan-1-amine hydrochloridyl
Molecular FormulaC17H18ClN2O
InChI KeyNIDXFJIFFPKQBY-UHFFFAOYSA-N
SMILES[Cl].NCCC1=CNC2=CC=C(OCC3=CC=CC=C3)C=C12
さらに表示
Infrared spectrumConforms
Appearance (Color)White to yellow to beige
Titration Argentometric>=97.5 %
HPLC>=97.5 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only