Methylthymol blue, tetrasodium salt, pure, water soluble
Methylthymol blue, tetrasodium salt, pure, water soluble
Methylthymol blue, tetrasodium salt, pure, water soluble
Thermo Scientific Chemicals

Methylthymol blue, tetrasodium salt, pure, water soluble

CAS: 1945-77-3 | C37H44N2Na4O13S | 848.779 g/mol
Have Questions?
Catalog NumberQuantity
1063700505 g
Catalog number 106370050
Price (EUR)
128,65
Online exclusive
143,00
Save 14,35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
128,65
Online exclusive
143,00
Save 14,35 (10%)
Each
Chemical Identifiers
CAS1945-77-3
IUPAC Nametetrasodium 2-[({3-[3-(3-{[bis(carboxylatomethyl)amino]methyl}-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-6-hydroxy-2-methyl-5-(propan-2-yl)phenyl}methyl)(carboxylatomethyl)amino]acetate
Molecular FormulaC37H40N2Na4O13S
InChI KeyDIZZDZCUMBBRSG-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Na+].[Na+].CC(C)C1=CC(=C(C)C(CN(CC([O-])=O)CC([O-])=O)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C(CN(CC([O-])=O)CC([O-])=O)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Specific absorbance A (1%/1cm)>=120 (at lambda max, in water)
Appearance (Color)Dark green to red-brown to black
Loss on drying=<10 % (110°C, 1 h)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only