Crystalline powder or crystals or crystalline mass
Melting point
37°C to 41°C
Appearance (Color)
White to light yellow to beige
Infrared spectrum
Conforms
GC
>=98.5 %
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.