1-Bromo-2-naphthol, 98%
1-Bromo-2-naphthol, 98%
1-Bromo-2-naphthol, 98%
Thermo Scientific Chemicals

1-Bromo-2-naphthol, 98%

CAS: 573-97-7 | C10H7BrO | 223.07 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 107161000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS573-97-7
IUPAC Name1-bromonaphthalen-2-ol
Molecular FormulaC10H7BrO
InChI KeyFQJZPYXGPYJJIH-UHFFFAOYSA-N
SMILESOC1=CC=C2C=CC=CC2=C1Br
さらに表示
GC>=97.5 %
Appearance (Color)Off-white to beige to brown-purple
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point77°C to 84°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only