Intact Virus Precipitation Reagent
Intact Virus Precipitation Reagent
Intact Virus Precipitation Reagent
Invitrogen™

Intact Virus Precipitation Reagent

Intact Virus Precipitation Reagent enables fast and efficient enrichment of intact, live viruses from dilute cell culture and virus transport자세히 알아보기
Have Questions?
카탈로그 번호수량
10720D100 reactions
카탈로그 번호 10720D
제품 가격(KRW)
605,000
온라인 행사
Ends: 30-Sep-2026
691,000
할인액 86,000 (12%)
Each
수량:
100 reactions
제품 가격(KRW)
605,000
온라인 행사
Ends: 30-Sep-2026
691,000
할인액 86,000 (12%)
Each

Intact Virus Precipitation Reagent enables fast and efficient enrichment of intact, live viruses from dilute cell culture and virus transport media. The enrichment is gentle and fast and can be performed in less than three hours. By tying up water molecules, the reagent forces less-soluble components such as vesicles out of solution, allowing them to be collected by short, low-speed centrifugation.

Intact Virus Precipitation Reagent can enrich for a plethora of viruses such as, but not limited to, SARS-CoV-2 coronavirus, respiratory syncytial virus (RSV), and Ebola-, Zika-, influenza A- viruses. The isolated intact virus can be used for functional studies.

Features of Intact Virus Precipitation Reagent include:
• Simple—3-step enrichment requiring only a centrifuge
• Fast—enrichment in <3 hours with minimal hands-on time
• Flexible volume—dilute virus samples can be enriched from large sample volumes
• Straight to PCR—no further nucleic acid extraction needed prior to PCR

Downstream applications
Enriched intact virus can be used for further virus growth or in any downstream functional assays, protein research (western blot), purification of RNA for PCR, etc. Virus isolated using the Intact Virus Precipitation Reagent requires no further nucleic acid extraction prior to doing PCR.

In addition, virus-like particles (VLPs), containing only virus-specific proteins and not nucleic acids, are extensively used in viral proteomic research. Intact Virus Precipitation Reagent isolates both live and inactivated intact viruses and VLPs.

Easy and efficient protocol
The Intact Virus Precipitation Reagent is simply added to the virus sample and incubate for two hours followed by a 30-minute centrifugation step. Remove the supernatant and keep the pellet with the enriched virus that is ready for any downstream analysis.

Automation ready with the KingFisher Purification System
For higher throughput automation needs, use the magnetic bead-based Dynabeads Intact Virus Enrichment strong anion exchange beads using any of our KingFisher Sample Purification Systems or liquid handlers.

Commercial supply
Our manufacturing sites are ISO 13485-certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility making them an ideal choice for commercial supply. If you are in the process of customizing Intact Virus Precipitation Reagent in a commercial product or service or need a larger bulk volume of this product, please contact us at oemdynal@lifetech.com or read more on our Dynabeads OEM page.

For research use only. Not for use in diagnostic procedures.
사양
용도(애플리케이션)Precipitation of viruses by tying up water molecules in the sample
용도(장비)Centrifuge capable of running upto 10,000 x G
분리 기술Precipitation by centrifugation
정제 타겟Viruses
수량100 reactions
유통 기한48 months from date of manufacture
배송 조건Ambient Temperature
제품 유형Intact Virus Precipitation Reagent
Unit SizeEach
구성 및 보관
Store at 2°C to 8°C.