n-Butyl acetate, 99+%, extra pure
n-Butyl acetate, 99+%, extra pure
n-Butyl acetate, 99+%, extra pure
Thermo Scientific Chemicals

n-Butyl acetate, 99+%, extra pure

CAS: 123-86-4 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 107750010
Price (USD)
69.65
Online Exclusive
77.40
Save 7.75 (10%)
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
69.65
Online Exclusive
77.40
Save 7.75 (10%)
Each
Chemical Identifiers
CAS123-86-4
IUPAC Namebutyl acetate
Molecular FormulaC6H12O2
InChI KeyDKPFZGUDAPQIHT-UHFFFAOYSA-N
SMILESCCCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Water=<0.1 % (K.F.)
Appearance (Color)Clear colorless
Refractive index1.3930 to 1.3950 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only