tert-Butylacetyl chloride, 99%
tert-Butylacetyl chloride, 99%
tert-Butylacetyl chloride, 99%
Thermo Scientific Chemicals

tert-Butylacetyl chloride, 99%

CAS: 7065-46-5 | C6H11ClO | 134.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 107790100
Price (USD)
35.65
Special offer
Online exclusive
Ends: 28-Dec-2026
41.30
Save 5.65
Each
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
35.65
Special offer
Online exclusive
Ends: 28-Dec-2026
41.30
Save 5.65
Each
Chemical Identifiers
CAS7065-46-5
IUPAC Name3,3-dimethylbutanoyl chloride
Molecular FormulaC6H11ClO
InChI KeyBUTKIHRNYUEGKB-UHFFFAOYSA-N
SMILESCC(C)(C)CC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration RCOOH=<1 %
Appearance (Form)Liquid
Titration RCOCl>=98.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only