3-Carbethoxy-4-piperidone hydrochloride, 98%
3-Carbethoxy-4-piperidone hydrochloride, 98%
3-Carbethoxy-4-piperidone hydrochloride, 98%
Thermo Scientific Chemicals

3-Carbethoxy-4-piperidone hydrochloride, 98%

CAS: 4644-61-5 | C8H13NO3·HCl | 207.66 g/mol
Have Questions?
Catalog NumberQuantity
108331000100 g
Catalog number 108331000
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS4644-61-5
IUPAC Nameethyl 4-oxopiperidine-3-carboxylate hydrochloridyl
Molecular FormulaC8H13ClNO3
InChI KeyOTNOTUGMYUCNQB-UHFFFAOYNA-N
SMILES[Cl].CCOC(=O)C1CNCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point169°C to 175°C
Appearance (Form)Crystalline powder
Appearance (Color)White to yellow
Infrared spectrumConforms
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only