2-chloroaniline, 98+%
2-chloroaniline, 98+%
2-chloroaniline, 98+%
Thermo Scientific Chemicals

2-chloroaniline, 98+%

CAS: 95-51-2 | C6H6ClN | 127.57 g/mol
数量:
250 mL
1 L
5 L
パッケージ:
すべて表示
Plastic drum
製品番号(カタログ番号) 108570050
価格(JPY)
-
数量:
5 L
パッケージ:
Plastic drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS95-51-2
IUPAC Name2-chloroaniline
Molecular FormulaC6H6ClN
InChI KeyAKCRQHGQIJBRMN-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1Cl
さらに表示
Appearance (Form)Liquid
GC>=98.0 %
AppearanceMay darken on storage
Appearance (Color)Clear light yellow to brown
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only