1-(4-Chlorobenzhydryl)piperazine, 90-95%
1-(4-Chlorobenzhydryl)piperazine, 90-95%
1-(4-Chlorobenzhydryl)piperazine, 90-95%
Thermo Scientific Chemicals

1-(4-Chlorobenzhydryl)piperazine, 90-95%

CAS: 303-26-4 | C17H19ClN2 | 286.79 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 108651000
Price (USD)
645.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
645.00
Each
Chemical Identifiers
CAS303-26-4
IUPAC Namehydrogen 1-[(4-chlorophenyl)(phenyl)methyl]piperazine chloride
Molecular FormulaC17H20Cl2N2
InChI KeyXFIUZNUONMJUIH-UHFFFAOYNA-N
SMILES[H+].[Cl-].ClC1=CC=C(C=C1)C(N1CCNCC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HClO4>=90.0 % (On dry substance)
TLC>=90 %
Appearance (Color)White to light beige
Melting point65°C to 75°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only