2-Chlorobenzonitrile, 99%
2-Chlorobenzonitrile, 99%
2-Chlorobenzonitrile, 99%
Thermo Scientific Chemicals

2-Chlorobenzonitrile, 99%

CAS: 873-32-5 | C7H4ClN | 137.57 g/mol
製品番号(カタログ番号) 108670010
価格(JPY)
-
数量:
1 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS873-32-5
IUPAC Name2-chlorobenzonitrile
Molecular FormulaC7H4ClN
InChI KeyNHWQMJMIYICNBP-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1C#N
さらに表示
Appearance (Form)Crystalline powder or crystals and/or
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)chunks or solid
Melting point43°C to 46°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only