Sf-900™ II SFM
Gibco™

Sf-900™ II SFM

Sf-900™ II SFMは、Spodoptera frugiperda(Sf9およびSf21)細胞の増殖と維持、また、バキュロウイルス発現ベクター系(BEVS詳細を見る
製品番号(カタログ番号)数量
109020881000 mL
10902096500 mL
109021046 x 1 L
109021615 L
1090217910 L
1090218720 L
製品番号(カタログ番号) 10902088
価格(JPY)
12,700
Each
数量:
1000 mL
Customize this product
Sf-900™ II SFMは、Spodoptera frugiperda(Sf9およびSf21)細胞の増殖と維持、また、バキュロウイルス発現ベクター系(BEVS)を用いて発現させる組換えタンパク質の大量産生に最適化された、無血清、無タンパク質の昆虫細胞培養培地です。この培地は、浮遊培養と単層培養のどちらにも適応し、他の鱗翅目の細胞増殖にも対応します。Sf-900™ II SFMの特長:

•長期・高密度培養に優れた性能を発揮
•組換えタンパク質産生用に最適化
•無血清、無タンパク質、そのまま使用可能な培地
•バイオリアクター培養にスケールアップ可能

長期・高密度培養に優れた性能を発揮
Sf-900™ II SFMで増殖されたSpodoptera frugiperda(Sf9)細胞は、9~12 x 106細胞/mLの最大細胞密度を達成し、他社製培地やグレース培地よりも大幅に改善されています(製品マニュアルを参照)。Lymantria dispar(マイマイガ)およびTrichoplusia ni(Tn-368、イラクサギンウワバ)細胞株でも、最大細胞密度の20~100%の増加が観察されています。Sf-900™ II SFMは、20継代を超える培養に対応します。

組換えタンパク質産生用に最適化
従来の組換えタンパク質の発現には、10% FBSを添加したグレース培地が使用されてきました。Sf-900™ II SFMは、Sf9および他の鱗翅目の細胞増殖、昆虫ウイルスおよびrDNAタンパク質の産生用に改良された、無血清、無タンパク質の昆虫細胞培養培地です。

無血清、無タンパク質、そのまま使用可能な培地
SF-900™ II SFMは、無血清の無タンパク質培地であるため、目的のタンパク質を簡単に精製できます。Sf-900™ II SFMはそのまま使用可能です。血清、グルタミン、界面活性剤を添加する必要はありません。他の市販の無血清培地に順応させた細胞は、通常、それ以上の順応を行わずに、直接Sf-900™ II SFMに継代できます。通常、細胞は血清含有培地からの順応が必要です。

バイオリアクター培養にスケールアップ可能
大規模細胞培養システムにおけるSf-900™ II SFMの有用性は、5 LのCelligen™ バイオリアクターで実証されています。rAcNPVによる感染を行い、rβ-GalおよびrEPOが生成されました(製品マニュアルを参照)。

製品の使用
製造プロセスでGibco™ Sf-900 II SFMをしており、FDAに申請を提出するお客様は、当社のタイプIIドラッグマスターファイル(DMF)を参照するために、当社に閲覧許可状を請求できます。
cGMP準拠の製造および品質管理システム
サプライチェーン継続性のため、Sf-900 II SFMは、ニューヨーク州グランドアイランドと英国スコットランドにある2施設で製造されています。どちらもcGMP製造要件に準拠した施設であり、ISO 13485認証を取得済みで、FDAに医療機器製造業者として登録されています。
研究用にのみ使用できます。診断用には使用いただけません。
仕様
細胞株Sf21, Sf9
細胞タイプInsect Cell
製品ラインGibco, Sf-900
製品タイプInsect Cell Serum Free Medium (SFM)
数量1000 mL
出荷条件Room Temperature
種S. frugiperda, Spodoptera frugiperda
分類Protein-free, Serum-free
形状Liquid
Serum LevelSerum-free
添加剤ありGlutamine
Unit SizeEach
組成および保存条件
Storage conditions: 2°C to 8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

よくあるご質問(FAQ)

Can Sf9 cells in Sf-900 III SFM (Cat. No. 12659017) be thawed and grown in Sf-900 II SFM instead?

It should be okay to thaw the cells into Sf-900 II SFM. This is a richer media compared to the Sf-900 III SFM so the cells would have an easy time adapting. We would recommend taking the cells through 3 passages in the new medium before using them for any experiments as that they have enough time to adapt.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What antimicrobials can be used in insect culture and at what concentration?

Many antibiotics are suitable for use with insect cells. The following antibiotics are commonly used:
- Penicillin/Streptomycine: 50-100 U/mL; 50-100 µg/mL
- Amphotericin B (Fungizone antimycotic): 0.25 µg/mL
- Gentamicin: 0.5 mL of 10 mg/mL solution in 500 mL media (final concentration: 10 µg/mL)

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is it necessary to heat-inactivate my serum before adding it to my medium?

Heat inactivation is not necessary. Our team has routinely used serum that has not been heat-inactivated, and we have not observed any effect on cell growth or morphology.

Many cells do not require heat-inactivated FBS. Some cells prefer heat-inactivated FBS. For instance, we use heat-inactivated FBS for our insect cell lines, i.e., Sf9 and Sf21 cells.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do you offer serum-free insect media?

Yes, we offer a variety of serum-free insect media. Please go here (http://www.thermofisher.com/us/en/home/life-science/cell-culture/insect-cell-culture/insect-cell-culture-misc/serum-free-media.html?icid=cvc-insect-media-c2t2) to view the media we offer and the differences between them.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

引用および参考文献 (4)

引用および参考文献
Abstract
Enhancing yield of infectious Bursal disease virus structural proteins in baculovirus expression systems: focus on media, protease inhibitors, and dissolved oxygen.
Authors: Hu Y C; Bentley W E;
Journal:Biotechnol Prog
PubMed ID:10585191
'Structural proteins of the poultry pathogen, infectious bursal disease virus (IBDV), were expressed in the baculovirus/insect cell expression system. To date, several reports have indicated that animal virus structural proteins are expressed only at low yield in this system. In this article, several factors were examined to enhance yield. These ... More
Titration of non-occluded baculovirus using a cell viability assay.
Authors:Mena JA, Ramírez OT, Palomares LA,
Journal:Biotechniques
PubMed ID:12613247
Enhanced growth of Sf-9 cells to a maximum density of 5.2 x 10(7) cells per mL and production of beta-galactosidase at high cell density by fed batch culture.
Authors:Elias CB, Zeiser A, Bédard C, Kamen AA,
Journal:Biotechnol Bioeng
PubMed ID:10745206
Significant improvement in cell growth and protein production has been achieved in Sf-9 insect cell cultures using pulse additions of multicomponent nutrient feed concentrates (Bédard et al., 1994; Chan et al., 1998). The present work focuses on investigating an alternative feeding strategy wherein the nutrients are fed in a semi ... More
Studies on the Mode of Ku Interaction with DNA.
Authors: Arosio Daniele; Cui Sheng; Ortega Claudia; Chovanec Miroslav; Di Marco Stefania; Baldini Giancarlo; Falaschi Arturo; Vindigni Alessandro;
Journal:J Biol Chem
PubMed ID:11796732
The Ku heterodimer plays a central role in non-homologous end-joining. The binding of recombinant Ku to DNA has been investigated by dynamic light scattering, double-filter binding, fluorescence spectroscopy, and band shift assays. The hydrodynamic radius of Ku in solution is 5.2 nm and does not change when a 25-bp double-strand ... More