Ciclobutanona, 98 %, Thermo Scientific Chemicals
Ciclobutanona, 98 %, Thermo Scientific Chemicals
Ciclobutanona, 98 %, Thermo Scientific Chemicals
Ciclobutanona, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ciclobutanona, 98 %, Thermo Scientific Chemicals

CAS: 1191-95-3 | C4H6O | 70.09 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
25 g
Envase:
Frasco de vidrio
Número de catálogo 110930250
Precio (USD)
-
Cantidad:
25 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS1191-95-3
EspecificacionesSpecification SheetHoja de especificaciones
Refractive index1.4200 to 1.4230 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only