Cyclododecanone, 99+%, Thermo Scientific Chemicals
Cyclododecanone, 99+%, Thermo Scientific Chemicals
Cyclododecanone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclododecanone, 99+%, Thermo Scientific Chemicals

CAS: 830-13-7 | Molecular Formula: C12H22O | Molecular Weight: 182.31 g/mol
Have Questions?
Catalog NumberQuantity
110975000500 g
Catalog number 110975000
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS830-13-7
IUPAC Namecyclododecanone
Molecular FormulaC12H22O
InChI KeySXVPOSFURRDKBO-UHFFFAOYSA-N
SMILESO=C1CCCCCCCCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point59°C to 61°C
GC>=99.0 %
Appearance (Form)Crystalline mass or crystals
Appearance (Color)White
Infrared spectrumConforms

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only