MEM Amino Acids Solution (50X) - FAQs
View additional product information for MEM Amino Acids Solution (50X) - FAQs (11130051)
2 product FAQs found
What is the solvent used for MEM Amino Acids Solution (50X)?
The solvent is distilled water.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is Gibco MEM Amino Acids Solution?
The MEM Amino Acids Solution is used as a growth supplement for cell culture medium, to help increase cell growth and viability. It is formulated to contain 50X the essential amino acids (except L-glutamine) found in the standard MEM. Find the complete formulation here (http://www.thermofisher.com/us/en/home/technical-resources/media-formulation.164.html).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.