Search
View additional product information for MEM Amino Acids Solution (50X) - FAQs (11130077)
1 product FAQs found
The solvent is distilled water.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.