Cyclohexanemethanol, 99%
Cyclohexanemethanol, 99%
Cyclohexanemethanol, 99%
Thermo Scientific Chemicals

Cyclohexanemethanol, 99%

CAS: 100-49-2 | C7H14O | 114.19 g/mol
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 111341000
Price (JPY)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS100-49-2
IUPAC Namecyclohexylmethanol
Molecular FormulaC7H14O
InChI KeyVSSAZBXXNIABDN-UHFFFAOYSA-N
SMILESOCC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only