Cystamine Dihydrochloride, 97%
Cystamine Dihydrochloride, 97%
Cystamine Dihydrochloride, 97%
Thermo Scientific Chemicals

Cystamine Dihydrochloride, 97%

CAS: 56-17-7 | C4H12N2S2·2HCl | 225.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 111775000
Price (EUR)
530,00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
530,00
Each
Chemical Identifiers
CAS56-17-7
IUPAC Namedihydrogen 2-[(2-aminoethyl)disulfanyl]ethan-1-amine dichloride
Molecular FormulaC4H14Cl2N2S2
InChI KeyYUFRRMZSSPQMOS-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NCCSSCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
Titration Argentometric>=96.0 %
Appearance (Form)Crystalline powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only