Dehydroacetic acid, 98%
Dehydroacetic acid, 98%
Dehydroacetic acid, 98%
Thermo Scientific Chemicals

Dehydroacetic acid, 98%

CAS: 520-45-6 | C8H8O4 | 168.15 g/mol
数量:
100 g
500 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 111945000
価格(JPY)
-
数量:
500 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS520-45-6
IUPAC Name3-acetyl-6-methyl-3,4-dihydro-2H-pyran-2,4-dione
Molecular FormulaC8H8O4
InChI KeyPGRHXDWITVMQBC-UHFFFAOYNA-N
SMILESCC(=O)C1C(=O)OC(C)=CC1=O
さらに表示
Sulfated ash=<0.1 % (700°C, 2 h)
Melting point109°C to 112°C
Appearance (Color)Cream to light yellow
Infrared spectrumConforms
Titration with NaOH>=97.5 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only