2,6-Dichloropurine, 97%
2,6-Dichloropurine, 97%
2,6-Dichloropurine, 97%
Thermo Scientific Chemicals

2,6-Dichloropurine, 97%

CAS: 5451-40-1 | C5H2Cl2N4 | 189.00 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1137000101 gGlass bottle
Catalog number 113700010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5451-40-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=96.0 %
Appearance (Color)White to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only