Dicyandiamide, 99.5%
Dicyandiamide, 99.5%
Dicyandiamide, 99.5%
Acros Organics

Dicyandiamide, 99.5%

CAS: 461-58-5 | C2H4N4 | 84.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
25 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number 113810025
also known as 11381
HSN 29312000
Price (INR)
16,400.00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
16,400.00
Each
Chemical Identifiers
CAS461-58-5
IUPAC NameN''-cyanoguanidine
Molecular FormulaC2H4N4
InChI KeyQGBSISYHAICWAH-UHFFFAOYSA-N
SMILESNC(N)=NC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<0.1 %
Melting point208°C to 211°C
Water=<0.5 % (K.F.)
Appearance (Color)White
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only