N,N'-Dicyclohexylcarbodiimide, 99%
N,N'-Dicyclohexylcarbodiimide, 99%
N,N'-Dicyclohexylcarbodiimide, 99%
Thermo Scientific Chemicals

N,N'-Dicyclohexylcarbodiimide, 99%

N,N-Dicyclohexylcarbodiimide, 99%, C13H22N2, CAS Number-538-75-0, carbodiimide, dicyclohexyl, bis cyclohexyl carbodiimide, dcc, 1,3-dicyclohexylcarbodiimide, carbodicyclohexylimide, n,n'-dicyclohexylcarbodiimide, n,n-dicyclohexylcarbodiimide, dcci, dicyclohexylcarbodiimide, dccd, 1kg | CAS: 538-75-0 | C13H22N2 | 206.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
5 kg
25 kg
Packaging:
View all
Plastic drum
Catalog number 113900050
Price (USD)
-
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS538-75-0
IUPAC NameN,N'-dicyclohexylmethanediimine
Molecular FormulaC13H22N2
InChI KeyQOSSAOTZNIDXMA-UHFFFAOYSA-N
SMILESC1CCC(CC1)N=C=NC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline mass or waxy solid
Infrared spectrumConforms
Melting point33°C to 35°C
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only