7-Diethylamino-4-methylcoumarin, 99%
7-Diethylamino-4-methylcoumarin, 99%
7-Diethylamino-4-methylcoumarin, 99%
Thermo Scientific Chemicals

7-Diethylamino-4-methylcoumarin, 99%

CAS: 91-44-1 | C14H17NO2 | 231.29 g/mol
数量:
5 g
100 g
500 g
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 114051000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS91-44-1
IUPAC Name7-(diethylamino)-4-methyl-2H-chromen-2-one
Molecular FormulaC14H17NO2
InChI KeyAFYCEAFSNDLKSX-UHFFFAOYSA-N
SMILESCCN(CC)C1=CC=C2C(C)=CC(=O)OC2=C1
さらに表示
Appearance (Color)Light beige to light purple
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline powder
Melting point70°C to 73°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only