Diethyl Ethoxymethylenemalonate, 99+%
Diethyl Ethoxymethylenemalonate, 99+%
Diethyl Ethoxymethylenemalonate, 99+%
Thermo Scientific Chemicals

Diethyl Ethoxymethylenemalonate, 99+%

CAS: 87-13-8 | C10H16O5 | 216.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 114345000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS87-13-8
IUPAC Name1,3-diethyl 2-(ethoxymethylidene)propanedioate
Molecular FormulaC10H16O5
InChI KeyLTMHNWPUDSTBKD-UHFFFAOYSA-N
SMILESCCOC=C(C(=O)OCC)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Refractive index1.4610 to 1.4640 (20°C, 589 nm)
Infrared spectrumConforms
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only