Diethyl phthalate, 99%
Diethyl phthalate, 99%
Diethyl phthalate, 99%
Thermo Scientific Chemicals

Diethyl phthalate, 99%

CAS: 84-66-2 | C12H14O4 | 222.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 114520025
Price (USD)
131.65
Online exclusive
146.00
Save 14.35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
131.65
Online exclusive
146.00
Save 14.35
Each
Chemical Identifiers
CAS84-66-2
IUPAC Name1,2-diethyl benzene-1,2-dicarboxylate
Molecular FormulaC12H14O4
InChI KeyFLKPEMZONWLCSK-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=CC=C1C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<25 APHA
Infrared spectrumConforms
Water=<0.2 % (K.F.)
Appearance (Form)Clear liquid
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only