N,N-Diethyl-m-toluamide, 98%
N,N-Diethyl-m-toluamide, 98%
N,N-Diethyl-m-toluamide, 98%
Thermo Scientific Chemicals

N,N-Diethyl-m-toluamide, 98%

CAS: 134-62-3 | C12H17NO | 191.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 114571000
Price (USD)
29.65
Online exclusive
32.30
Save 2.65
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
29.65
Online exclusive
32.30
Save 2.65
Each
Chemical Identifiers
CAS134-62-3
IUPAC NameN,N-diethyl-3-methylbenzamide
Molecular FormulaC12H17NO
InChI KeyMMOXZBCLCQITDF-UHFFFAOYSA-N
SMILESCCN(CC)C(=O)C1=CC=CC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<100 APHA
GC>=97.5 %
Infrared spectrumConforms
Water=<0.3 %
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only