4,6-Dihydroxy-2-mercaptopyrimidine, 98%
4,6-Dihydroxy-2-mercaptopyrimidine, 98%
4,6-Dihydroxy-2-mercaptopyrimidine, 98%
Thermo Scientific Chemicals

4,6-Dihydroxy-2-mercaptopyrimidine, 98%

CAS: 504-17-6 | C4H4N2O2S | 144.15 g/mol
数量:
5 g
100 g
500 g
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 114961000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS504-17-6
IUPAC Name2-sulfanylidene-1,3-diazinane-4,6-dione
Molecular FormulaC4H4N2O2S
InChI KeyRVBUGGBMJDPOST-UHFFFAOYSA-N
SMILESO=C1CC(=O)NC(=S)N1
さらに表示
Infrared spectrumConforms
SolubilityColorless to yellow to light brown solution
Titration with NaOH97.5 to 102.5 %
Solubility(5% in 1M NaOH) Clear to slightly hazy
Appearance (Form)Powder or crystalline solid
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only