2,3-Dihydroxynaphthalene, 97%
2,3-Dihydroxynaphthalene, 97%
2,3-Dihydroxynaphthalene, 97%
Thermo Scientific Chemicals

2,3-Dihydroxynaphthalene, 97%

CAS: 92-44-4 | C10H8O2 | 160.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number 115020500
Price (EUR)
45,00
Each
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
45,00
Each
Chemical Identifiers
CAS92-44-4
IUPAC Namenaphthalene-2,3-diol
Molecular FormulaC10H8O2
InChI KeyJRNGUTKWMSBIBF-UHFFFAOYSA-N
SMILESOC1=C(O)C=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Infrared spectrumConforms
Melting point161°C to 165°C
Appearance (Color)White to beige to pink to light grey
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only