3,5-Dimethoxy-4-hydroxycinnamic acid, 98%, predominantly trans isomer
3,5-Dimethoxy-4-hydroxycinnamic acid, 98%, predominantly trans isomer
3,5-Dimethoxy-4-hydroxycinnamic acid, 98%, predominantly trans isomer
Thermo Scientific Chemicals

3,5-Dimethoxy-4-hydroxycinnamic acid, 98%, predominantly trans isomer

CAS: 530-59-6 | C11H12O5 | 224.21 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
產品號碼 115550010
價格 (TWD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS530-59-6
IUPAC Name(2E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
Molecular FormulaC11H12O5
InChI KeyPCMORTLOPMLEFB-ONEGZZNKSA-N
SMILESCOC1=CC(C=CC(O)=O)=CC(OC)=C1O
檢視更多
規格Specification Sheet規格表
Appearance (Color)Yellow to light beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only