N,N'-Diphenylbenzidine, 97%
N,N'-Diphenylbenzidine, 97%
N,N'-Diphenylbenzidine, 97%
Thermo Scientific Chemicals

N,N'-Diphenylbenzidine, 97%

CAS: 531-91-9 | C24H20N2 | 336.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 117210100
Price (EUR)
85,70
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
85,70
Each
Chemical Identifiers
CAS531-91-9
IUPAC NameN4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
Molecular FormulaC24H20N2
InChI KeyFDRNXKXKFNHNCA-UHFFFAOYSA-N
SMILESN(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point246°C to 250°C
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Color)Grey to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only