Dithiooxamide, 98%
Dithiooxamide, 98%
Dithiooxamide, 98%
Thermo Scientific Chemicals

Dithiooxamide, 98%

CAS: 79-40-3 | C2H4N2S2 | 120.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 117550050
Price (USD)
92.65
Special offer
Online exclusive
Ends: 28-Dec-2026
109.00
Save 16.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
92.65
Special offer
Online exclusive
Ends: 28-Dec-2026
109.00
Save 16.35
Each
Chemical Identifiers
CAS79-40-3
IUPAC Nameethanedithioamide
Molecular FormulaC2H4N2S2
InChI KeyOAEGRYMCJYIXQT-UHFFFAOYSA-N
SMILESNC(=S)C(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red to brown
Appearance (Form)Powder
Infrared spectrumConforms
Elemental Analysis19.5 to 20.5 % (C)
Sulfated ash=<2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only