Esculin hydrate, 97%
Esculin hydrate, 97%
Esculin hydrate, 97%
Thermo Scientific Chemicals

Esculin hydrate, 97%

CAS: 531-75-9 | C15H16O9 | 340.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 117830050
Price (EUR)
46,65
Online exclusive
51,70
Save 5,05
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
46,65
Online exclusive
51,70
Save 5,05
Each
Chemical Identifiers
CAS531-75-9
IUPAC Name7-hydroxy-6-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2H-chromen-2-one
Molecular FormulaC15H16O9
InChI KeyXHCADAYNFIFUHF-TYKRLAFXNA-N
SMILESOC[C@H]1O[C@@H](OC2=C(O)C=C3OC(=O)C=CC3=C2)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation(on dry substance)
Infrared spectrumConforms
Titration with NaOH>=96.0 % (On dry substance)
Specific optical rotation-84° to -87° (20°C, 589 nm) (c=1.5, dioxane/H2O 1:1)
Appearance (Color)White to cream
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only